Documents
GESE25010A75
GE SE250 mini-vertical unit for two slab gels
complete, gel size 10 cm × 8 cm, AC/DC input 500 V AC
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6147-45
Cytiva SE250-10A-,75
gel size
10 cm × 8 cm
General description
Runs two standard 10×8 cm mini-gels. The upper buffer chamber is also an efficient heat exchanger for active temperature control. The unit can run 12 tube gels with two tube gel adapter kits.