Skip to Content
MilliporeSigma
All Photos(1)

Documents

GERPN5661

CyDye Post-Labelling Reactive Dye Pack

Cytiva RPN5661

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.31

manufacturer/tradename

Cytiva RPN5661

packaging

pkg of 1pack = 12 x Cy3 + 12 x Cy5

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Specifically optimized for nucleic acid microarray labeling. Higher reactivity Cy3 and Cy5 dyes: 40 000 pmol reactive dye per vial, at >75% reactive dye content. Ready-to-use, individually dispensed, and packed in foil packs.

CyDye Post-Labeling Reactive Dye Pack for the generation of highly fluorescent Cy3- and Cy5-labeled probes via post-labeling (amino allyl) method. The only reactive CyDye optimized for microarray probe labelling. CyDye Post-Labelling Reactive Dye Pack provides conveniently packaged, highly reactive Cy3 and Cy5 monofunctional NHS ester to label probes via the post-labeling (amino allyl) route for use in microarray applications.

Features and Benefits

  • The only reactive CyDye fluors specifically optimized for nucleic acid microarray labeling.
  • Higher reactivity Cy3 and Cy5 dyes: 40 000 pmol reactive dye per vial, at >75% reactive dye content.
  • Ready-to-use, individually dispensed, and packed in foil packs for protective storage.
  • Use of these reagents avoids waste of excess dye and the need to aliquot and store material from large pack sizes.
  • Includes 12 vials Cy3 NHS ester and 12 vials Cy5 NHS ester.
  • For the generation of highly fluorescent Cy3- and Cy5-labeled probes via post-labeling (amino allyl) method.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact LSlicensing@cytiva.com.

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy3 is a trademark of Cytiva
CyDye is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service