Skip to Content
MilliporeSigma
All Photos(1)

Documents

GERPN418

ECL Advance Blocking Reagent

Cytiva RPN418

Synonym(s):

ECL Blocking Agent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

feature

wetted part: no

packaging

pkg of 40 g

manufacturer/tradename

Cytiva RPN418

color

Yellow. Pale color

storage temp.

2-8°C

General description

During the Western blotting process, non-specific binding of antibodies to the membrane is detrimental to the specificity and sensitivity of the assay. To overcome this challenge, it is essential to block parts of the membrane not already occupied by proteins. The Amersham ECL Prime blocking agent can be used for blocking Amersham Protran nitrocellulose (NC) and Amersham Hybond polyvinylidene fluoride (PVDF) membranes when used along with the ECL™ chemiluminescence detection reagents, such as Amersham ECL™, ECL™ start, ECL™ Prime, ECL™ Select, and ECL™ Plex. Each pack contains 40 g of blocking agent, which is sufficient for at least 20 mini blots.

Application

ECL Advance Blocking Reagent may be used for high-sensitivity ECL and fluorescence western blotting applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service