Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE90-1002-73

Sigma-Aldrich

Superdex® 200 PG, XK 26/100,90- (24-44 μm) HPLC Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
SB.52

product name

Superdex® 200 PG, XK26/100,90-9,

material

glass column

product line

Superdex®

packaging

1 ea of

bed size

26 mm × 900 mm

bed volume

480 mL

column I.D.

26 mm

matrix

composite of cross-linked agarose and dextran
polymeric

particle size

24-44 μm

cleaning

1-14

working range

3-12

suitability

suitable for bioprocess medium

separation technique

size exclusion (SEC)

General description

HiLoad Superdex 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.
  • High resolution with short run times and good recovery.
  • High capacity at high flow rates.
  • Separations can be scaled up to production levels.
  • Prepacked for convenience and reproducibility.
  • 1/16” fittings for convenient connections to ÄKTA design systems.


Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex 30 pg), Mr 3 000 to 70 000 (Superdex 75 pg), and Mr 10 000 600 000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service