GE80-6204-26
Mini tank transfer unit, TE22
power consumption 50 W, AC/DC input 100 V AC, output500 mA
About This Item
Recommended Products
description
EPS2A201 suggested power supply
feature
wetted part: no
manufacturer/tradename
Cytiva 80620426
parameter
2 environmental limits (Pollution degree)
4-40 °C temp. range (Indoor use)
80% humidity limit (2000 m3/h)
W × H × D
14 cm × 24 cm × 16.5 cm
output
500 mA
power consumption
50 W
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service