Key Documents
GE80-1129-36
GelBond PAGfilm 124 X 258 mm, (Pkg/50)
Cytiva 80-1129-36, pack of 50 pieces
Select a Size
1 PKG
$654.00
$654.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$654.00
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
$654.00
Please contact Customer Service for Availability
packaging
pack of 50 pieces
manufacturer/tradename
Cytiva 80-1129-36
availability
not available in Japan
storage temp.
room temp
Related Categories
General description
GelBond PAGfilm is a polyester film with a treated surface which binds to the agarose gel.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)