Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE29268402

HiTrap® MabSelect SuRe LX

Cytiva 29268402, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NB.13

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 29268402

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
  • Reduces the risk of product contamination and carryover
  • Improves performance and reduces overall costs
  • High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes

Legal Information

HiTrap is a registered trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service