GE28-9513-78
Protein A Mag Sepharose™
Cytiva 28-9513-78, pack of 4 × 500 μL
Select a Size
$242.00
Select a Size
About This Item
$242.00
Recommended Products
ligand
native protein A
packaging
pack of 4 × 500 μL
manufacturer/tradename
Cytiva 28-9513-78
storage condition
(20% Ethanol, 20% medium slurry)
parameter
Room temperature temp. range
matrix
paramagnetic, spherical, highly cross-linked agarose particles
capacity
8- 17 mg binding capacity(human IgG/ml gel)
storage temp.
2-8°C
Related Categories
General description
Application
Features and Benefits
- Visible and dense Sepharose™ based magnetic beads with Protein A ligand, easy to spot and collect bound target Protein
- Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
- Simple capture of target Protein in small or large sample volumes (Low microliter to high milliliter scale)
- Optimized capacity for enrichment/immunoprecipitation of target Proteins - only Low amounts of antibody needed
- Flexible protocols with elution conditions optimized for both electrophoresis and MS analysis
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page shows various purification options for Protein A Sepharose chromatography media and describes typical binding and elution conditions for Protein A Sepharose chromatography media.
This page describes immunoprecipitation (immunoaffinity or pull-down techniques).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service