Recommended Products
manufacturer/tradename
Cytiva
General description
illustra plasmidPrep Midi Flow Kit can be used to process bacterial sample volumes of 25 to 50 ml for high copy number plasmid DNA and up to 500 ml for low copy number plasmid DNA. The kit consists of a high-performance, anion exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA. The kit contains fast-flow plasmid 250 columns, resuspension, lysis, neutralization, wash, elution, and TE buffers, and a full protocol booklet with a detachable, quick reference protocol card.
Features and Benefits
> 10 endotoxin units (EU) per μg of plasmid DNA.
> 70% (strain-dependent) of supercoiled pDNA.
Anion exchange chromatography.
High column capacity allows for a proportional yield for input sample volume of 25 to 50 ml (for high copy number plasmid) and up to 500 ml for low copy number plasmid.
Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns.
Modified alkaline lysis procedure.
Provides plasmid DNA with excellent purity and low endotoxin levels.
Purification format is single-use, gravity flow illustra Fast-Flow plasmid 250 columns.
Legal Information
illustra is a trademark of Cytiva
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service