Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9042-65

illustra Blood Mini , 250

Cytiva 28-9042-65

Synonym(s):

Blood Mini Kit, illustra Blood Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$883.00

$883.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$883.00

About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

$883.00


Please contact Customer Service for Availability

packaging

pkg of 250 ea

manufacturer/tradename

Cytiva 28-9042-65

capacity

60 μg binding capacity

Related Categories

General description

Blood genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from whole blood, buffy coat, bone marrow, and nucleated red blood cells.

Genomic DNA is found in the chromosomes and it is much longer and higher in molecular weight than plasmid DNA. Genomic DNA is the code for the genome, and this is what is needed to do genome sequencing.

For sequencing it is desirable that the genomic DNA extraction technique used allows the collection of intact genomic DNA in the longest fragments possible. A kit format can help standardize the procedures used to prepare tissue and cell genomic DNA samples for analysis.

The purification process involves minimal shearing resulting in the production of 5 to 10 μg of good quality, intact genomic DNA from a 200 μL sample. Input sample volumes can vary from 50 μL to 1 mL.

The method uses a simple genomic DNA purification protocol. This protcol that uses chaotropic agents to extract DNA from blood cells, denature protein components, and promote the selective binding of DNA to a column-based, novel silica membrane. This results in high molecular weight DNA that can be used downstream (e.g., in bone marrow genomic DNA for next generation sequencing).

The elution volume can be modified as necessary to obtain the appropriate genomic DNA concentration for most downstream molecular biological applications.

The genomic DNA kit contains spin columns prepacked with a novel silica membrane, lyophilized proteinase K powder, lysis solution, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.

Application

Suitable for use in molecular biology applications including standard, multiplex and quantitative PCR; cloning; haplotyping; restriction enzyme digestion; and genotyping.

Features and Benefits

  • High-quality genomic DNA extraction from blood with a greater proportion of higher molecular weight DNA and less shearing
  • Demonstrated application to various types of whole blood including human, horse, rabbit, rat, and mouse
  • Increased ease of use with reduced pipetting volume changes, one centrifugation setting, and color-coded kit components
  • Verified in several downstream applications including reverse transcription polymerase chain reaction (RT-PCR), endpoint PCR, multiplex PCR, and restriction enzyme digests

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows downstream applications after genomic DNA purification.

Description of common problems that can occur during genomic DNA preparation

Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.

Protocols

This page shows Genomic DNA purification using illustra™ genomicPrep Mini Spin Kits from Cytiva.

This page shows genomic DNA purification from sample on FTA elute from Cytiva.

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service