Key Documents
GE28-9034-66
illustra GFX PCR DNA and Gel Band Purification Kit, 10 purifications
About This Item
UNSPSC Code:
41106300
NACRES:
NA.52
Product GE28-9034-66 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Cytiva
General description
illustra GFX PCR DNA and Gel Band Purification Kit contains the following components in sufficient quantities: GFX columns, collection tubes, color-coded bottles of capture buffer, wash buffer, and two elution buffers (Tris-HCl and sterile water), and an instruction booklet.
Features and Benefits
Addition of a visual color indicator to the capture buffer ensures optimal pH for maximum DNA binding and recovery.
Color-coded caps for ease of use.
Fast and easy-to-use method with less than 10 min hands-on time.
Flexible 10 to 50 μl elution volume for different DNA concentration needs.
For the isolation and concentration of DNA fragments from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.
Highly pure DNA is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning.
Improved column design with increased maximum volume, holding of up to 800 μl liquid volume (950 μl without a cap).
Two choices of elution buffer provided to fit downstream applications.
Legal Information
illustra is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
This page shows the procedure for purifying and concentrating DNA using illustra™ GFX 96 PCR Purification Kit from Cytiva.
Protocols
DNA cleanup using illustra™ GFX kits from Cytiva.