Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-9263-01

Ready-to-go T-Primed First-Strand Kit

Cytiva 27-9263-01, sufficient for 55 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

usage

sufficient for 55 reactions

feature

hotstart: no

manufacturer/tradename

Cytiva 27-9263-01

color

colorless

General description

The kit provides 50 single-dose, ambient-temperature-stable, first-strand cDNA synthesis reactions, each containing NotI-(dT)18 Primer, Cloned FPLCpure M-MuLV Reverse Transcriptase, RNase Inhibitor, and nucleotides.

Application

The kit provides 50 single-dose, ambient-temperature-stable, first-strand cDNA synthesis reactions, each containing NotI-(dT)18 Primer, Cloned FPLCpure M-MuLV Reverse Transcriptase, RNase Inhibitor, and nucleotides. (A reaction mixture containing rabbit globin mRNA and specific globin primers is included as a control).

Features and Benefits

  • For synthesis of full-length first-strand cDNA templates from polyadenylated RNA for use in RT-PCR.
  • Preformulated, ambient-temperature-stable, single-dose glassified reaction mixes minimize pipetting errors, avoid potential cross-contamination of subsequent PCR products, and ensure optimal performance from each reaction.
  • Predispensed reaction mixes are highly suitable for research applications which use PCR to detect and quantify eukaryotic RNA species isolated from a variety of cell sources and tissue samples.
  • Reaction mixes include NotI-(dT)18 primer in sufficient quantity to serve as a downstream PCR primer for performing 3′-RACE. Final reaction volume is 33 μL once reconstituted.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ready-To-Go is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service