Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-1798-01

CY®3,5 Amidite, 100MG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

manufacturer/tradename

Cytiva

solubility

soluble (Soluble in cold water and hot water)

General description

For incorporation of Cy3.5 fluorescent dyes at any position in an oligonucleotide during synthesis.

Linkage of Cy dyes allows labeled DNA to hybridize to other DNA.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Other Notes

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use.

Legal Information

Cy is a registered trademark of Cytiva

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service