Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-1524-01

M13K07 Helper Phage

Cytiva 27-1524-01, pack of 100 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106501
NACRES:
NA.56

form

liquid

packaging

pack of 100 μL

manufacturer/tradename

Cytiva 27-1524-01

color

Colorless to light yellow

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

The phage is an M13mp1 derivative containing a modified gene 11[G changed to T at position 6,125, giving a met to ile change at codon 40 of the gene 11 protein (1)], an insertion of the origin of replication of p15A and the kanamycin resistance gene from Tn903 at the Ava I site (position 5,825).

Application

  • For phage rescue

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service