Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE19-5037-01

C Tubes

joint: 10/40, Cytiva

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NB.43

Joint

joint: 10/40

manufacturer/tradename

19-5037-01
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

Resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.

  • Operating Pressure Maximum: 1 bar (0.1 MPa ) (14.5 psi)
  • Operating Temperature: 4°C-60°C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service