Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17548911

HiTrap® Capto Lentil Lectin

pkg of 5 × 1 mL, Cytiva, 17-5489-11, 75 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21

packaging

pkg of 5 × 1 mL

manufacturer/tradename

17-5489-11
Cytiva

avg. part. size

75 μm

General description

Capto Lentil Lectin is an affinity chromatography resin for purification of glycoproteins and other molecules containing carbohydrates such as a-D-mannose and a-D-glucose or sterically related residues.
  • Group-specific adsorbent for carbohydrate-containing molecules
  • High productivity and cost-efficiency in downstream operations
  • Animal component-free production


Capto Lentil Lectin resin is based on a rigid high-flow agarose base matrix that enables high flow rates. The product is available in bulk as well as in various high-throughput process development formats.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service