Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5171-01

Mono P® 5/200 GL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

ligand

mixed quaternary and tertiary amines

description

Ion Exchanger Type (Chromatofocusing (weak anion exchanger))

packaging

pkg of 4 mL

parameter

580 psi

bed size

5 mm × 200 mm

bed volume

4 mL

column I.D.

5.7 mm

matrix

monodisperse polystyrene/divinyl benzene

matrix active group

amine phase

avg. part. size

10 μm

cleaning

2-14

working range

2-12

capacity

0.15-0.21 mmol Cl-/mL binding capacity

separation technique

anion exchange

Application

Mono P® is based on the same matrix as Mono Q® and Mono S. The 10 μm MonoBeads particles are substituted with tertiary and quaternary amines and the small bead size contributes significantly to the high resolution that can be achieved.

Features and Benefits

  • Greatly concentrates samples and resolves pI differences of 0.02 pH units.
  • Mono P® 5/50 GL for rapid scouting of elution conditions (pH gradients) or for separations not requiring highest resolution.
  • Mono P® 5/200 GL for highest resolution separation.
  • Monodisperse 10 μm porous beads yield high resolution, dynamic capacity, reproducibility, and durability.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Mono P is a registered trademark of Cytiva
Mono Q is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service