Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5168-01

Sigma-Aldrich

Mono S® 5/50 GL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

ligand

-OCH2CH(OH)CH2OCH2CH(OH)CH2SO3-

material

PEEK column

description

Ion Exchanger Type (Methyl sulfonate strong cation)

packaging

pkg of 1 mL

parameter

580 psi

bed size

5 mm × 50 mm

bed volume

1 mL

column I.D.

5.7 mm

matrix

monodisperse porous polystyrene/divinyl benzene beads
polymeric

matrix active group

methyl sulfonate phase

avg. part. size

10 μm

cleaning

1-14

working range

2-12

separation technique

cation exchange

General description

Mono S® is a strong cation exchanger prepacked with MonoBeads in a Tricorn column. The column is an excellent choice for small-scale polishing in purification of proteins, peptides, and other biomolecules when high purity is required. The column has 1 mL bed volume for small-scale purifications.

The Tricorn columns offer high-resolution separations with greater loading capacity but slightly lower resolution than MiniBeads columns.

Features and Benefits

  • Monodisperse 10 μm porous beads yield high resolution, dynamic capacity, reproducibility, and durability.
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers.
  • Compatible with AKTA design, FPLC System, and other high-performance LC systems.
  • MonoBeads are also available as Mono P® for chromatofocusing

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

FPLC is a trademark of Cytiva
Mono P is a registered trademark of Cytiva
Mono S is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page provides information about performing an isolation of recombinant protein complexes with different pull-down assays with products from Cytiva.

This page shows how to perform a purification of His-tagged membrane proteins.

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

See All

Protocols

This page covers using Monobeads for purification of proteins, peptides, or oligonucleotides.

This page shows how to condition membrane proteins for further analysis with products from Cytiva.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service