Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1279-02

rProtein A Sepharose Fast Flow

Cytiva 17-1279-02, pack of 25 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$3,260.00

$3,260.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$3,260.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

$3,260.00


Please contact Customer Service for Availability

ligand

recombinant protein A (E. coli)

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-1279-02

storage condition

(20% Ehtanol)

matrix

4% cross-linked agarose

particle size

60-165 μm

average diameter

90 μm (d50v)

cleaning in place

2-11

working range

3-10

capacity

~50 mg binding capacity (human lgG/ml medium)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

General description

rProtein A Sepharose 4 Fast Flow is an affinity resin for antibody purification at laboratory and process scale.

Monoclonal and polyclonal antibody purification

rProtein A Sepharose 4 Fast Flow is built upon the well-established Sepharose Fast Flow platform and is used in many approved monoclonal antibody processes. It is composed of recombinant protein A coupled to the cross-linked 4% agarose base matrix.The highly crosslinked agarose resin enables high flow rates and high productivity.

Protein A resin characteristics

The recombinant protein is produced in E. coli and engineered with an oriented coupling to enhance binding capacity. Epoxy-based coupling ensures low ligand leakage. Specificity of the recombinant protein for the Fc region of IgG is similar to the native protein, giving a high degree of selectivity and yield.

Streamlined scale up

Scale-up with this resin is straightforward and is further aided by a range of prepacked process development tools, making purification scale-up straightforward.

Features and Benefits

  • For monoclonal and polyclonal antibody purification at both laboratory and process scale.
  • Well established among Protein A resins as excellent for monoclonal antibody (mAb) processes.
  • Free from animal-derived components.
  • Hydrophilic base matrix ensures low levels of non-specific binding and low levels of host cell-derived impurities in the elution pool.
  • Fulfills industrial demands for security of supply, robust performance, and regulatory support.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service