Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE17-0854-02

NAP-10 50 ST

Cytiva 17-0854-02

Synonym(s):

NAP-10 Column, NAP-10 Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

packaging

pkg of 50 columns

manufacturer/tradename

Cytiva 17-0854-02

storage condition

(.01 M NaOH)

particle size

20-80 μm (dry)

working range

2-13

capacity

<1 mL binding capacity

General description

Disposable columns prepacked with Sephadex® G-25 DNA Grade and require only gravity to run.

NAP Columns are disposable columns prepacked with Sephadex® G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration, washing and sample elution.

Features and Benefits

  • For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flow.
  • Prepacked with Sephadex® G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide.
  • Available in three sizes depending on sample volume: 0.5 mL (NAP-5), 1 mL (NAP-10) or 2.5 mL (NAP-25).
  • Useful for small-scale purification, desalting and buffer exchange.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

NAP is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service