Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0448-02

Cytodex® 1 microcarrier beads

Cytiva, pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41121800
NACRES:
NC.01
Product GE17-0448-02 is not currently sold in your country. Contact Technical Service

material

dextran beads

sterility

non-sterile

packaging

pkg of 100 g

manufacturer/tradename

17-0448-02
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

A general-purpose microcarrier for most established, anchorage-dependent cell lines.

High yield culture of anchorage-dependent cells: Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few mL to more than 6000 L. Using Cytodex in simple suspension culture systems provides yields of several million cells per mL.

Suitable for most established cell lines: Formed by substituting a cross-linked dextran matrix with positively charged DEAE* groups distributed throughout the matrix, is a general purpose microcarrier. It is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.

Features and Benefits

  • General-purpose microcarrier for most established, anchorage-dependent cell lines.
  • Contains positive charges throughout the microporous matrix.
  • Transparent for easy microscopic examination of attached cells.

Preparation Note

Prior to use, swell beads in PBS (50 mL/g) and autoclave at 120 °C.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service