Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0042-02

Sephadex® G-50

Fine, Cytiva, pack of 500 g

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,260.00

$2,260.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$2,260.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115722
NACRES:
NA.56

$2,260.00


Please contact Customer Service for Availability

feature

autoclavable In wet form (pH 7.0) at 120°C for 30 min

packaging

pack of 500 g

manufacturer/tradename

Cytiva

parameter

<Darcy´s Law flow rate

swelling

1 g swells to 9-11 mL

particle size

20-80 μm (dry)
(wet)

cleaning in place

2-13

working range

2-10

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sephadex® G-50 Fine is well established gel filtration medium for desalting and buffer exchange of biomolecules >30 000 molecular weight. The Fine´s small bead size give higher efficiency.

Sephadex® is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-50 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-50 is available in 4 different particle sizes (Course, Medium, Fine & Superfine) and Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Cource and Medium are are preferred for large scale group separations at where high flow rates and low operating pressures are required.

Features and Benefits

  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
  • Suitable for purification of DNA from small molecules by gel filtration
  • Small bead size for shorter diffusion distances
  • Classic gel filtration medium.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service