Key Documents
GE11-0002-93
Ettan spot picker non-backed gel kit
Synonym(s):
11-0002-93, protein handling
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
manufacturer/tradename
Cytiva 11-0002-93
Cytiva 11000293 (Article Number)
General description
  • Allows quick and accurate protein spot picking from nonbacked 2-D gels, increasing the versatility of Ettan Spot Picker.
  • Compatible with common staining methods such as silver, Coomassie Blue, fluorescence, and labeling with Ettan DIGE Fluor dyes.
  • High-precision picking with reference markers - using the gel frame, a nonbacked gel slab is attached to a temporary cellophane backing, supporting the reference markers.
  • Improves nonbacked gel handling during scanning, and the glass lid protects the moist gel.
Kit Contains:
  • 1 x Picking platform
  • 1 x Gel frame (outer frame and inner frame)
  • 1 x Gel loading platform
  • 50 x Porous cellophane
  • 2 x Glass lid for gel carrier
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)