Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE10600065

Amersham Protran® Premium Western blotting membranes, nitrocellulose

pore size 0.45 μm, sheet W × L 140 mm × 160 mm, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 25 ea

manufacturer/tradename

Cytiva 10600065

sheet W × L

140 mm × 160 mm

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Nitrocellulose blotting membrane, provides excellent sensitivity, resolution, and low background in fluorescence Western blotting detection applications.

Application

Amersham Protran® Premium Western blotting membranes, nitrocellulose may be used in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • High binding capacity (162 – 180 µg Iµg/cm2).
  • Suitable for proteins of a wide range of molecular weights.
  • Unsupported, 100% pure nitrocellulose membrane for use with all ECL Western blotting systems and protein blotting applications.
  • Available in popular precut sizes for laboratory application.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service