Key Documents
GBL505384
Grace Bio-Labs ONCYTE® NOVA nitrocellulose film slides
for 384 well SBS compliant microtiter plates/glass substrates, square pads, L × W 2.5 mm × 2.5 mm
Synonym(s):
hybridization chambers, incubation chambers, nitrocellulose slides, protein microarrays
Select a Size
1 EA
$365.00
$365.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$365.00
About This Item
UNSPSC Code:
41122600
NACRES:
NB.15
$365.00
Please contact Customer Service for Availability
material
nitrocellulose (on glass)
packaging
pack of 1 ea
manufacturer/tradename
Grace Bio-Labs 505384
L × W
2.5 mm × 2.5 mm
binding type
medium binding surface
General description
Grace Bio-Labs ONCYTE® NOVA nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Legal Information
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)