Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9294

Sigma-Aldrich

Anti-GABAA Receptor (α5 subunit), Cytosolic Loop antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-γ-Aminobutyric Acid Type A Receptor (α5 subunit)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

usage

sufficient for 10 blots

mol wt

antigen apparent mol wt 55 kDa

species reactivity

rat

technique(s)

immunoprecipitation (IP): 20 μg using 150 μL dodecylsulfate extracts of rat forebrains
western blot: 1:1,000 using rat forebrain

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

Immunogen

fusion protein of MBP with the amino acid sequence representing the cytosolic loop of the rat GABAA receptor (α5 subunit)

Target description

GABAA Receptor (α5 subunit), Cytosolic Loop is an ionotropic receptor that forms the GABA gated chloride channel and consists of several heterogeneous subunits with membrane recognition sites for benzodiazepines.

Physical form

Solution in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 ug/ml BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service