Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9262

Sigma-Aldrich

Gly-Pro 4-methoxy-β-naphthylamide hydrochloride

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O3 · HCl
CAS Number:
Molecular Weight:
363.84
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

form

powder

color

white to off-white

solubility

methanol: 50 mg/mL, clear, light yellow

storage temp.

−20°C

SMILES string

Cl.COc1cc(NC(=O)C2CCCN2C(=O)CN)cc3ccccc13

InChI

1S/C18H21N3O3.ClH/c1-24-16-10-13(9-12-5-2-3-6-14(12)16)20-18(23)15-7-4-8-21(15)17(22)11-19;/h2-3,5-6,9-10,15H,4,7-8,11,19H2,1H3,(H,20,23);1H

InChI key

IYLMARVOIYCGET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Substrate for dipeptidyl aminopeptidase IV

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gossrau et al.
Histochemistry, 80(2), 183-186 (1984-01-01)
The lymphocytes of the rat thymus can be grossly differentiated by their cell membrane-bound proteinases. Subcapsular thymocytes lack aminopeptidase A (APA) and AMP and gamma-glutamyltranspeptidase (GGT). Cortical thymocytes show a high activity of APA but no APM and no GGT.
Studies on dipeptidyl(amino)peptidase IV (glycyl-proline naphthylamidase). II. Blood vessels.
Z Lojda
Histochemistry, 59(3), 153-166 (1979-01-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service