Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8773

Sigma-Aldrich

Glycerol

suitable for electrophoresis, ≥99%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

698 °F

technique(s)

electrophoresis: suitable

impurities

≤0.005% Chlorinated compounds (as Cl)

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

heavy metals (as Pb): ≤5 ppm

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc J A Stevens et al.
Microbial cell factories, 10, 61-61 (2011-08-05)
Lactobacillus reuteri metabolizes glycerol to 3-hydroxypropionaldehyde (3-HPA) and further to 1,3-propanediol (1,3-PDO), the latter step catalysed by a propanediol dehydrogenase (PDH). The last step in this pathway regenerates NAD+ and enables therefore the energetically more favourable production of acetate over
Timothy J Kidd et al.
Journal of clinical microbiology, 47(5), 1503-1509 (2009-03-06)
Pseudomonas aeruginosa is an important cause of pulmonary infection in cystic fibrosis (CF). Its correct identification ensures effective patient management and infection control strategies. However, little is known about how often CF sputum isolates are falsely identified as P. aeruginosa.
David Baddeley et al.
PloS one, 6(5), e20645-e20645 (2011-06-10)
Optical super-resolution imaging of fluorescently stained biological samples is rapidly becoming an important tool to investigate protein distribution at the molecular scale. It is therefore important to develop practical super-resolution methods that allow capturing the full three-dimensional nature of biological
Diego Pasini et al.
Genes & development, 22(10), 1345-1355 (2008-05-17)
Polycomb group (PcG) proteins regulate important cellular processes such as embryogenesis, cell proliferation, and stem cell self-renewal through the transcriptional repression of genes determining cell fate decisions. The Polycomb-Repressive Complex 2 (PRC2) is highly conserved during evolution, and its intrinsic
V Quisth et al.
Diabetologia, 48(5), 946-953 (2005-03-22)
The regulation of skeletal muscle lipolysis is not fully understood. In the present study, the effects of systemic and local noradrenaline administration on lipolysis and blood flow rates in skeletal muscle and adipose tissue were studied in vivo. First, circulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service