Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8660

Sigma-Aldrich

Gastrointestinal Tumor Antigen from human fluids

≥50,000 units/mL, buffered aqueous solution

Synonym(s):

Cancer-associated, Ca 19-9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

biological source

human fluids

form

buffered aqueous solution

concentration

≥50,000 units/mL

storage temp.

−20°C

Quality

Highly purified

Unit Definition

One unit is an arbitrary amount related to a reference antigen preparation using CIS-RIA method.

Physical form

Solution in 0.15 M phosphate buffered saline, pH 7.4, containing 0.1% sodium azide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service