Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8293

Sigma-Aldrich

Galanin (2-11) porcine, rat

≥97% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$733.00

$733.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
$733.00

About This Item

Empirical Formula (Hill Notation):
C54H81N13O14
Molecular Weight:
1136.30
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

$733.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](N)Cc2c[nH]c3ccccc23)[C@@H](C)O)C(N)=O

InChI

1S/C54H81N13O14/c1-26(2)17-37(46(57)73)62-49(76)38(18-27(3)4)63-51(78)40(20-31-13-15-33(70)16-14-31)61-44(72)24-59-47(74)29(7)60-53(80)42(25-68)66-52(79)41(22-43(56)71)64-50(77)39(19-28(5)6)65-54(81)45(30(8)69)67-48(75)35(55)21-32-23-58-36-12-10-9-11-34(32)36/h9-16,23,26-30,35,37-42,45,58,68-70H,17-22,24-25,55H2,1-8H3,(H2,56,71)(H2,57,73)(H,59,74)(H,60,80)(H,61,72)(H,62,76)(H,63,78)(H,64,77)(H,65,81)(H,66,79)(H,67,75)/t29-,30+,35-,37-,38-,39-,40-,41-,42-,45-/m0/s1

InChI key

SUJPPMXJCZETOA-ZKEPBWIVSA-N

Gene Information

rat ... Gal(29141)

Amino Acid Sequence

Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-NH2

General description

Galanin (2-11) is a galanin receptor type 2 agonist.[1][2]

Biochem/physiol Actions

Studies in mice show that injection of galanin (2-11) has anticonvulsant activity in seizure affected animals with an intact DR (dorsal root)–hippocampal serotonergic profile, and has no effect in animals with absent 5-HT (hydroxytryptamine) profile.[1]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey M Mazarati et al.
Journal of neurochemistry, 95(5), 1495-1503 (2005-10-13)
The neuropeptide galanin suppresses seizure activity in the hippocampus by inhibiting glutamatergic neurotransmission. Galanin may also modulate limbic seizures through interaction with other neurotransmitters in neuronal populations that project to the hippocampus. We examined the role of galanin receptors types
Xiaoying Lu et al.
Neuropeptides, 39(3), 165-167 (2005-06-10)
The neuropeptide galanin regulates a variety of physiological and pathophysiological processes through three G protein coupled receptors, GalR1, GalR2, and GalR3. The studies on galanin receptor subtype specific effects have been hampered by the lack of high affinity subtype selective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service