Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8272

Sigma-Aldrich

Galanin rat

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C141H211N43O41
CAS Number:
Molecular Weight:
3164.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(C)NC(=O)C(Cc1cnc[nH]1)NC(=O)C2CCCN2C(=O)CNC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(Cc3ccc(O)cc3)NC(=O)CNC(=O)C(C)NC(=O)C(CO)NC(=O)C(CC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(NC(=O)C(Cc4c[nH]c5ccccc45)NC(=O)CN)C(C)O)C(=O)NC(CC(O)=O)C(=O)NC(CC(N)=O)C(=O)NC(Cc6cnc[nH]6)C(=O)NC(CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(Cc7ccccc7)C(=O)NC(CO)C(=O)NC(CC(O)=O)C(=O)NC(CCCCN)C(=O)NC(Cc8cnc[nH]8)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(C(C)O)C(N)=O

InChI

1S/C141H211N43O41/c1-15-71(10)113(139(224)177-100(52-112(199)200)132(217)173-97(49-105(144)191)129(214)171-96(48-81-57-151-66-159-81)128(213)166-86(30-23-37-152-141(147)148)122(207)178-102(62-186)136(221)169-92(43-76-25-17-16-18-26-76)127(212)180-103(63-187)137(222)174-99(51-111(197)198)131(216)165-85(29-21-22-36-142)121(206)170-94(46-79-55-149-64-157-79)120(205)155-59-109(195)163-88(40-68(4)5)133(218)182-114(74(13)188)116(146)201)181-118(203)73(12)161-123(208)95(47-80-56-150-65-158-80)175-138(223)104-31-24-38-184(104)110(196)60-156-119(204)87(39-67(2)3)167-124(209)89(41-69(6)7)168-126(211)91(44-77-32-34-82(190)35-33-77)164-108(194)58-154-117(202)72(11)160-135(220)101(61-185)179-130(215)98(50-106(145)192)172-125(210)90(42-70(8)9)176-140(225)115(75(14)189)183-134(219)93(162-107(193)53-143)45-78-54-153-84-28-20-19-27-83(78)84/h16-20,25-28,32-35,54-57,64-75,85-104,113-115,153,185-190H,15,21-24,29-31,36-53,58-63,142-143H2,1-14H3,(H2,144,191)(H2,145,192)(H2,146,201)(H,149,157)(H,150,158)(H,151,159)(H,154,202)(H,155,205)(H,156,204)(H,160,220)(H,161,208)(H,162,193)(H,163,195)(H,164,194)(H,165,216)(H,166,213)(H,167,209)(H,168,211)(H,169,221)(H,170,206)(H,171,214)(H,172,210)(H,173,217)(H,174,222)(H,175,223)(H,176,225)(H,177,224)(H,178,207)(H,179,215)(H,180,212)(H,181,203)(H,182,218)(H,183,219)(H,197,198)(H,199,200)(H4,147,148,152)

InChI key

JHQDYHHNJBOXKP-UHFFFAOYSA-N

Gene Information

rat ... Gal(29141)

Amino Acid Sequence

Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile-Asp-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-His-Gly-Leu-Thr-NH2

Biochem/physiol Actions

Co-localized with acetylcholine and inhibits its release; inhibits the release of glutamic acid, decreases excitability of spinal neurons; blocks voltage-activated Ca2+ channels; linked to behavioral and cognitive deficits in Alzheimer′s disease.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Korolkiewicz et al.
Fundamental & clinical pharmacology, 11(6), 576-583 (1997-01-01)
Porcine galanin (1-29)-NH2, galantide (M15) and galanin (1-14)-(alpha-aminobutyric acid8)-scyliorhinin-I used in concentrations of 300, 1,000 and 3,000 nM respectively caused contractions of rat fundus strips. The contractile responses to galanin(1-29)-NH2 were not modified by atropine (10 microM), guanethidine (10 microM)
S Wang et al.
FEBS letters, 434(3), 277-282 (1998-09-22)
The neuropeptide galanin mediates a diverse array of physiological functions through activation of specific receptors. Roles of the three recently cloned galanin receptors (GalRs) in rat intestinal contraction and food intake were examined using GalR-selective ligands and the results were
J F Todd et al.
Endocrinology, 139(10), 4222-4229 (1998-09-29)
Recent evidence suggests that pituitary galanin synthesized in the lactotroph is a paracrine regulator of lactotroph proliferation and PRL secretion and that these effects are mediated via a pituitary-specific galanin receptor, GAL-R2(orig.). At this receptor subtype, the galanin fragment 3-29
Rachel P Tillage et al.
Brain structure & function, 225(2), 785-803 (2020-02-18)
Accumulating evidence indicates that disruption of galanin signaling is associated with neuropsychiatric disease, but the precise functions of this neuropeptide remain largely unresolved due to lack of tools for experimentally disrupting its transmission in a cell type-specific manner. To examine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service