Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

G7522

Sigma-Aldrich

3-α-Galactobiose

synthetic, ≥90%

Synonym(s):

α-D-Gal-(1→3)-D-Gal, 3-O-α-D-Galactopyranosyl-D-galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
83008
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

Assay

≥90%

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@@H]2[C@@H](O)C(O)O[C@H](CO)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gisella L Puga Yung et al.
Xenotransplantation, 19(3), 196-206 (2012-06-19)
Anti-Galα1,3Galβ-R natural antibodies are responsible for hyperacute rejection in pig-to-primate xenotransplantation. Although the generation of pigs lacking the α1,3galactosyltransferase (GalT) has overcome hyperacute rejection, antibody-mediated rejection is still a problem. It is possible that other enzymes synthesize antigens similar to
T Biedermann et al.
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete, 63 Suppl 1, 76-79 (2012-05-11)
Delayed immediate-type allergy to innards and red meat can be mediated by IgE antibodies to galactose-α-1, 3-galactose (α-Gal). Apart from humans and Old World apes, α-Gal is ubiquitously expressed in glycoproteins and glycolipids. Thus, as α-Gal is immunogenic for humans
Delayed urticaria to beef ingestion.
Henry Benjamin Coker et al.
Journal of the American Academy of Dermatology, 68(3), e101-e101 (2013-02-12)
Beatrice Lyimo et al.
Bioscience, biotechnology, and biochemistry, 76(2), 336-342 (2012-02-09)
A galactose specific lectin (CpL) was purified from the Clavaria purpurea mushroom by affinity chromatography. CpL agglutinated only trypsin-treated rabbit erythrocytes. On enzyme linked lectin sorbent assay (ELLSA), the lectin bound with thyroglobulin and asialo bovine submaxillary mucin (BSM). The
Darius Ghaderi et al.
Biotechnology & genetic engineering reviews, 28, 147-175 (2012-05-24)
One of the fastest growing fields in the pharmaceutical industry is the market for therapeutic glycoproteins. Today, these molecules play a major role in the treatment of various diseases, and include several protein classes, i.e., clotting factors, hormones, cytokines, antisera

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service