Documents
G6266
Sigma-Aldrich
Guards for Sigma-Aldrich® pH electrodes
For 120 mm stem length
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
General description
Removable plastic guard that protects the glass bulb from impact during use.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany