Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5787

Sigma-Aldrich

L-Glutamic acid-[3,4-3H]

≥95% (Radiochemical Purity), aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (Radiochemical Purity)

form

aqueous ethanol solution

extent of labeling

30-80 Ci/mmol

mass shift

M+8

storage temp.

2-8°C

SMILES string

[3H]C([3H])([C@H](N)C(O)=O)C([3H])([3H])C(O)=O

Packaging

serum bottle

Physical form

Aqueous solution containing 2% ethanol

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service