Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G4795

Sigma-Aldrich

Anti-GLUT10 (SLC2A10) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Glucose Transporter 10, Anti-Solute Carrier Family 2 (Facilitated Glucose Transporter) Member 10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa by calculation

species reactivity

human

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SLC2A10(81031)

Immunogen

peptide corresponding to the human GLUT10 protein (amino acids 367-385).

Biochem/physiol Actions

GLUT10 is a glucose transporter that is encoded by the gene SLC2A10, and is highly expressed in liver and pancreas. Polymorphisms in SLC2A10 may not influence the pathogenesis of type 2 diabetes but the expression of GLUT10 and the interaction with TGF-β signaling pathway has important role in the development of peripheral arterial disease in patients with type 2 diabetes. GLUT10 is also required for the development of cardiovascular system and the notochord in zebrafish embryos.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gitte Andersen et al.
Diabetes, 52(9), 2445-2448 (2003-08-28)
The SLC2A10 gene encodes the GLUT10 facilitative glucose transporter, which is expressed in high amounts in liver and pancreas. The gene is mapped to chromosome 20q12-q13.1, a region that has been shown to be linked to type 2 diabetes. The
Andy Willaert et al.
Human molecular genetics, 21(6), 1248-1259 (2011-11-26)
Growth factor signaling results in dramatic phenotypic changes in cells, which require commensurate alterations in cellular metabolism. Mutations in SLC2A10/GLUT10, a member of the facilitative glucose transporter family, are associated with altered transforming growth factor-β (TGFβ) signaling in patients with
Yi-Der Jiang et al.
BMC medical genetics, 11, 126-126 (2010-08-26)
Recent data indicate that loss-of-function mutation in the gene encoding the facilitative glucose transporter GLUT10 (SLC2A10) causes arterial tortuosity syndrome via upregulation of the TGF-β pathway in the arterial wall, a mechanism possibly causing vascular changes in diabetes. We genotyped

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service