Skip to Content
MilliporeSigma
All Photos(3)

Documents

G4173

Sigma-Aldrich

Anti-GLUT4 antibody produced in rabbit

1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

GLUT4 Antibody - Anti-GLUT4 antibody produced in rabbit, Glut4 Antibody, Anti-Glucose transporter 4, Anti-SLC2A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ≥58 kDa

species reactivity

human, mouse

concentration

1.5 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using C2C12 cells
western blot: 1-2 μg/mL using C2C12 cell lysate and HepG2 cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SLC2A4(6517)
mouse ... Slc2a4(20528)
rat ... Slc2a4(25139)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Elodie Cattin et al.
The Journal of biological chemistry, 290(44), 26699-26714 (2015-09-12)
Aging and diseases generally result from tissue inability to maintain homeostasis through adaptation. The adult heart is particularly vulnerable to disequilibrium in homeostasis because its regenerative abilities are limited. Here, we report that MLIP (muscle enriched A-type lamin-interacting protein), a
Yue Guan et al.
BioMed research international, 2015, 457246-457246 (2015-05-09)
Impaired glucose tolerance (IGT), known as the prediabetes stage, is usually induced by habits of life or environmental factors. Established IGT animal models are mostly conducted with chemical compounds such as streptozocin or genetic modification. However, the occasion of exposure
Michalina Alicka et al.
Stem cell research & therapy, 11(1), 4-4 (2020-01-05)
Progressive loss of cell functionality caused by an age-related impairment in cell metabolism concerns not only mature specialized cells but also its progenitors, which significantly reduces their regenerative potential. Adipose-derived stem cells (ASCs) are most commonly used in veterinary medicine
Wenying Ren et al.
Adipocyte, 2(1), 17-28 (2013-04-20)
Protein palmitoylation, by modulating the dynamic interaction between protein and cellular membrane, is involved in a wide range of biological processes, including protein trafficking, sorting, sub-membrane partitioning, protein-protein interaction and cell signaling. To explore the role of protein palmitoylation in
Marianna Di Chiara et al.
American journal of physiology. Renal physiology, 309(9), F779-F790 (2015-09-04)
The Rab GTPase-activating protein TBC1D4 (AS160) controls trafficking of the glucose transporter GLUT4 in adipocytes and skeletal muscle cells. TBC1D4 is also highly abundant in the renal distal tubule, although its role in this tubule is so far unknown. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service