Skip to Content
MilliporeSigma
All Photos(1)

Documents

G3545

Sigma-Aldrich

GW583340 dihydrochloride

≥97% (HPLC), solid

Synonym(s):

4-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenylamino]-6-[2-[[2-(methylsulfonyl)ethyl]aminomethyl]-4-thiazolyl]quinazoline dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H25ClFN5O3S2 · 2HCl
CAS Number:
Molecular Weight:
671.03
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

mp

259.8-262.4 °C (lit.)

solubility

DMSO: 9 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].CS(=O)(=O)CCNCc1nc(cs1)-c2ccc3ncnc(Nc4ccc(OCc5cccc(F)c5)c(Cl)c4)c3c2

InChI

1S/C28H25ClFN5O3S2.2ClH/c1-40(36,37)10-9-31-14-27-35-25(16-39-27)19-5-7-24-22(12-19)28(33-17-32-24)34-21-6-8-26(23(29)13-21)38-15-18-3-2-4-20(30)11-18;;/h2-8,11-13,16-17,31H,9-10,14-15H2,1H3,(H,32,33,34);2*1H

InChI key

WIMITXDBYLKRKB-UHFFFAOYSA-N

Biochem/physiol Actions

Potent dual ErbB-2/EGFR tyrosine kinase inhibitor. IC50= 0.01 and 0.014 μM for EGFR and ErbB-2. Orally active.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service