Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

G3516

Sigma-Aldrich

α-D-Glucoheptonic acid sodium salt

Synonym(s):

D-Glycero-D-guloheptonic acid, Glucosemonocarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H13O8Na
CAS Number:
Molecular Weight:
248.16
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99% (HPLC)

form

powder

optical activity

[α]20/D 5.5 to 6.5 °, c = 4% (w/v) in water

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)C(O)C(O)C([O-])=O

InChI

1S/C7H14O8.Na/c8-1-2(9)3(10)4(11)5(12)6(13)7(14)15;/h2-6,8-13H,1H2,(H,14,15);/q;+1/p-1/t2-,3-,4+,5-,6-;/m1./s1

InChI key

FMYOMWCQJXWGEN-WYRLRVFGSA-M

Biochem/physiol Actions

α-D-Glucoheptonic acid has been shown to inhibit germ-tube formation in C. albicans budding yeast cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Pollack et al.
Journal of general microbiology, 133(2), 415-424 (1987-02-01)
It has been reported that Candida albicans can form germ-tubes only in the narrow pH range of 6-8, and that by changing only the pH one can regulate germ-tube formation. We found that the pH minimum for germ-tube formation could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service