Skip to Content
MilliporeSigma
All Photos(1)

Documents

G2543

Sigma-Aldrich

Geranylgeranyl monophosphate ammonium salt solution

≥95% (TLC), methanol:ammonia solution

Synonym(s):

all trans-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraenyl monophosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35O4P · 2NH3
Molecular Weight:
404.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (TLC)

form

methanol:ammonia solution

packaging

vial of 200 μg

shipped in

wet ice

storage temp.

−20°C

SMILES string

[NH4+].[NH4+].C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\COP([O-])([O-])=O

InChI

1S/C20H35O4P.2H3N/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-24-25(21,22)23;;/h9,11,13,15H,6-8,10,12,14,16H2,1-5H3,(H2,21,22,23);2*1H3/b18-11+,19-13+,20-15+;;

InChI key

FNVCAIOHPLGDJG-CNKFFNGESA-N

Other Notes

Possibly involved in isoprenoid metabolism

Physical form

Solution in methanol:10 mM ammonium hydroxide (7:3)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service