Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G2000

Sigma-Aldrich

2-(β-D-Galactosidoxy)naphthol AS-LC

Synonym(s):

2-(β-D-Galactosidoxy)naphthalene-3-carboxylic acid (2,5-dimethoxy-4-chloroanilide, Naphthol AS-LC β-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26ClNO9
CAS Number:
Molecular Weight:
519.93
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

storage temp.

−20°C

SMILES string

COc1cc(NC(=O)c2cc3ccccc3cc2OC4OC(CO)C(O)C(O)C4O)c(OC)cc1Cl

InChI

1S/C25H26ClNO9/c1-33-18-10-16(19(34-2)9-15(18)26)27-24(32)14-7-12-5-3-4-6-13(12)8-17(14)35-25-23(31)22(30)21(29)20(11-28)36-25/h3-10,20-23,25,28-31H,11H2,1-2H3,(H,27,32)

InChI key

UJPWFVGXQNDAFV-UHFFFAOYSA-N

Substrates

Substrate for the histochemical demonstration of galactosidase.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruilin Zhang et al.
Food technology and biotechnology, 58(4), 455-464 (2021-01-29)
Non-alcoholic steatohepatitis is a potentially progressive hepatic disorder that can lead to end-stage liver disease and hepatocellular carcinoma. The inhibitory effects of proteins and hydrolysates from the liver of newborn piglets on hepatic steatosis in oleic acid-induced hepatocellular carcinoma (HepG2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service