Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G1376

Sigma-Aldrich

L-Glutamic acid ammonium salt

≥99% (TLC)

Synonym(s):

(S)-2-Aminopentanedioic acid ammonium salt, Glu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8NO4 · NH4
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (TLC)

SMILES string

N.NC(CCC(O)=O)C(O)=O

Biochem/physiol Actions

L-Glutamic acid is a natural α proteinogenic amino acid use in a wide range of research application including use as a cell culture supplement, enzyme substrate or receptor ligand. L-Glutamine is an agonist at kainate, NMDA, and quisqualate glutamatergic receptors and an excitatory amino acid neurotransmitter.
Agonist at kainate, NMDA, and quisqualate glutamatergic receptors; an excitatory amino acid neurotransmitter.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Flores-Soto et al.
Neurologia (Barcelona, Spain), 27(5), 301-310 (2012-01-06)
To review the physiology of the glutamate receptor subunits such as N-methyl-D-aspartate (NMDA). Glutamic acid (Glu) is the major excitatory neurotransmitter in the central nervous system which interacts with two types classified into two types: metabotropic and ionotropic. Ionotropic receptors
K Sugiyama et al.
Neuroscience, 32(3), 779-791 (1989-01-01)
Several reports have described apparently normal survival and development of hippocampal and spinal cord culture preparations grown in Ham's F-12 medium, which contains 100 microM each of L-glutamate and L-aspartate. As at this concentration these amino acids are neurotoxic, some
Sandra Stoppelkamp et al.
Journal of neuroscience methods, 194(1), 132-138 (2010-10-19)
The cultivation of pure neuronal cultures is considered advantageous for the investigation of cell-type specific responses (such as transmitter release and also pharmacological agents), however, divergent results are a likely consequence of media modifications and culture composition. Using Fura-2 based
Jennifer C M Lau et al.
Molecular vision, 19, 1538-1553 (2013-07-24)
This study investigated changes in the transcript levels of genes related to glutamate neurotransmission and transport as diabetes progresses in the Long-Evans rat retina. Transcript levels of vascular endothelial growth factor (VEGF), erythropoietin, and insulin-like growth factor binding protein 3
V V Kolobov et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (1)(1), 27-32 (2013-06-29)
In experiments on rats showed that intranasal administration of glutamate antibodies in a dose of 300 microg/kg after 1 h after bilateral injection of neurotoxic fragment of beta-amyloid protein (25-35)--Abeta(25-35)--into the Meynert nuclei restores learning ability in the test of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service