G115060
DMT-dG(dmf) Phosphoramidite
configured for ABI
Synonym(s):
DMT-dG(dmf) Amidite, N-[(dimethylamino)methylene]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N2-Dimethylformamidine-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]
Select a Size
Select a Size
About This Item
Recommended Products
type
for DNA synthesis
Quality Level
product line
Proligo Reagents
Assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder
technique(s)
oligo synthesis: suitable
color
white to off-white
λ
conforms (UV/VIS Identity)
compatibility
configured for ABI
nucleoside profile
base: deoxyguanosine
base protecting group: DMF
2' protecting group: none
5' protecting group: DMT
deprotection: fast/standard
storage temp.
-10 to -25°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(\N=C/N(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6
InChI
1S/C43H53N8O7P/c1-29(2)51(30(3)4)59(56-24-12-23-44)58-36-25-38(50-28-45-39-40(50)47-42(48-41(39)52)46-27-49(5)6)57-37(36)26-55-43(31-13-10-9-11-14-31,32-15-19-34(53-7)20-16-32)33-17-21-35(54-8)22-18-33/h9-11,13-22,27-30,36-38H,12,24-26H2,1-8H3,(H,47,48,52)/b46-27-/t36-,37+,38+,59?/m0/s1
InChI key
YRQAXTCBMPFGAN-UJASEYITSA-N
Looking for similar products? Visit Product Comparison Guide
General description
- dG(dmf) is deprotected faster than the conventional dG(ib): thedeprotection time in concentrated ammonia is reduced to 2 hours at55 °C or 1 hour at 65 °C
- The dG(dmf)-monomer is especially suitable for G-rich sequences:incomplete deprotection is greatly reduced in comparison with theconventional dG(ib)-monomer
- dG(dmf)-phosphoramidite is as stable in solution as the standard dA(bz)-,dC(bz)- and dT-phosphoramidites
- dG(dmf)-phosphoramidite can directly substitute for dG(ib)-phosphoramidite
- No change is required in the reagents commonly used for DNA synthesis(except a low concentration iodine oxidizer i.e., 0.02 M in iodine, should beemployed)DMT-dG(dmf) Phosphoramidite is configured for ABI Synthesizers."
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service