Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G114N0-HH

Sigma-Aldrich

NPPOC-dG(ipac) RayDite

Synonym(s):

NPPOC-dG(ipac) Amidite, NPPOC-dG(ipac) Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H51N8O11P
Molecular Weight:
850.85
UNSPSC Code:
41116105

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

Assay

≥96% (31P-NMR)
≥96% (31P-NMR)
≥96.0% (reversed phase HPLC)
≥96.0% (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for H-NMR
conforms to structure for NMR (13C-NMR)
conforms to structure for NMR (13C-NMR)

shipped in

dry ice

storage temp.

−20°C
−20°C

InChI

1S/C40H51N8O11P/c1-24(2)28-13-15-29(16-14-28)54-22-34(49)43-39-44-37-36(38(50)45-39)42-23-46(37)35-19-32(59-60(57-18-10-17-41)47(25(3)4)26(5)6)33(58-35)21-56-40(51)55-20-27(7)30-11-8-9-12-31(30)48(52)53/h8-9,11-16,23-27,32-33,35H,10,18-22H2,1-7H3,(H2,43,44,45,49,50)/t27?,32-,33+,35+,60?/m0/s1

InChI key

ZHHHLRWGITWEAU-DWRAGFEYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

This product and it use are covered under US Patent Number 5,763,599
RayDite is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service