Skip to Content
MilliporeSigma
All Photos(2)

Documents

G0800

Sigma-Aldrich

GS21 Supplement (50X)

Serum-free Neural Media Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

description

Designed to be used in conjunction with N3100.

sterility

sterile; sterile-filtered

form

liquid

shelf life

4 weeks (at 2-8 °C after thaw)

concentration

50 X (Stock Concentration)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

GS21 Supplement is a serum-free neural media supplement designed to improve the overall perfomance of the primary neurons. GS21 is specifically optimized for the propagation and long-term viability of primary rat and mouse neurons in culture, without the need for co-culture with astrocyte feeder cells.

Application

Primary rat and mouse neuronal cell culture.

Features and Benefits

  • Significantly improves long-term viability of rat and mouse primary neurons in vitro.
  • Enhances neurite outgrowth of primary neurons in culture.
  • Supports growth and maintenance of primary neurons at low or high cell density plating.

Preparation Note

Product Dilution:

1. GS21 Supplement is supplied as a 50X concentrate. Dilute 2mL in 100mL of your neuronal basal media.
2. For primary cortical neuron cultures, supplement with 0.5 mM dipeptide glutamine before use.
3. Once combined with media, store at 2 °C to 8 °C in the dark for up to 4 weeks.

Legal Information

GS21 is a trademark of GlobalStem

supplement

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroscientists claim growing pains.
Daniel Cressey
Nature, 459(7243), 19-19 (2009-05-09)
Yucui Chen et al.
Journal of neuroscience methods, 171(2), 239-247 (2008-05-13)
In vitro culturing of primary neurons is a mainstay of neurobiological research. Many of these culture paradigms have taken advantage of defined culture media rather than serum additives that contain undefined survival factors to facilitate experimental manipulations and interpretation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service