G0752
Gly-Asp dihydrate
Synonym(s):
Glycyl-L-aspartic acid dihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
storage temp.
−20°C
SMILES string
O.NCC(=O)N[C@@H](CC(O)=O)C(O)=O
InChI
1S/C6H10N2O5.H2O/c7-2-4(9)8-3(6(12)13)1-5(10)11;/h3H,1-2,7H2,(H,8,9)(H,10,11)(H,12,13);1H2/t3-;/m0./s1
InChI key
SKIXLVGMUJDROR-DFWYDOINSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 1), o11-o13 (2007-01-09)
The title bis(glycyl-L-aspartic acid) oxalate complex {systematic name: bis[2-(2-ammonioacetamido)butanedioic acid] oxalate 0.4-hydrate}, 2C6H11N2O5+.C2O4(2-).4H2O, crystallizes in a triclinic space group with the planar peptide unit in a trans conformation. The asymmetric unit consists of two glycyl-L-aspartic acid molecules with positively charged
Journal of inorganic biochemistry, 77(3-4), 135-139 (2000-03-04)
The first solution studies at physiological pH for the formation of metal complexes of taurine, +NH3CH2CH2S03-, one of the most abundant low molecular weight organic compounds in the animal kingdom, are reported. The complexes Cu(Gly-GlyH-1) (1) and [Cu(Gly-AspH-1)] (2) react
Journal of neurochemistry, 56(1), 129-135 (1991-01-01)
Canavan disease, an autosomal recessive disorder, is characterized biochemically by N-acetylaspartic aciduria and aspartoacylase (N-acyl-L-aspartate amidohydrolase; EC 3.5.1.15) deficiency. However, the role of aspartoacylase and N-acetylaspartic acid in brain metabolism is unknown. Aspartoacylase has been purified to apparent homogeneity with
International journal of peptide and protein research, 19(2), 206-211 (1982-02-01)
The crystal structure of the acidic dipeptide glycyl-L-aspartic acid dihydrate, Gly-L-Asp X 2H2O, C6H10N2O5 X 2H2O, has been determined by means of three-dimensional counter X-ray data. The dipeptide crystallizes in space group P212121 of the orthorhombic system with four formula
Biochimica et biophysica acta, 1770(8), 1212-1218 (2007-06-19)
To better understand the mechanism of vanadium reduction in ascidians, we examined the reduction of vanadium(V) to vanadium(IV) by NADPH and the reduction of vanadium(IV) to vanadium(III) by L-cysteine methyl ester (CysME). UV-vis and electron paramagnetic resonance spectroscopic studies indicated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service