Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0541

Sigma-Aldrich

Anti-Glutamate Receptor NMDAR1 (NR1) (Splice Variant Insert N1) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen 100 kDa

species reactivity

rat

packaging

antibody small pack of 25 μg

technique(s)

dot blot: 1:1000
immunohistochemistry: 1:1000
western blot: 1:1000

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

synthetic peptide representing the sequence of the N1 splice variant.

Application

Anti-Glutamate Receptor NMDAR1 (NR1) (Splice Variant Insert N1) antibody is suitable for dot blot, immunohistochemistry and western blot at a dilution of 1:1000.

Biochem/physiol Actions

NMDA receptors (NMDAR) are glutamate-gated ion channels. It is sensitive to N-methyl-D-aspartate (NMDA) and consists of several homologous subunits (NMDAR1). NMDAR is highly expressed in the central nervous system and involved in excitatory synaptic transmission as well as synaptic plasticity. The activity of NMDA receptors are modulated by glycine. It exhibits high calcium permeability and unique voltage-dependent sensitivity to magnesium. It has been reported that phosphorylated NMDA receptors may regulate their function and play a role in many forms of synaptic plasticity such as long-term potentiation.

Physical form

Lyophilized powder from 5 mM ammonium bicarbonate (some residual salt may be present)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Dong Xia et al.
Asian journal of andrology, 20(4), 355-359 (2018-03-09)
Differences in intravaginal ejaculation latency reflect normal biological variation, but the causes are poorly understood. Here, we investigated whether variation in ejaculation latency in an experimental rat model is related to altered sympathetic nervous system (SNS) activity and expression of
Jie Yang et al.
Andrology, 9(1), 352-360 (2020-08-05)
Estimated 30%-40% of patients with chronic prostatitis/chronic pelvic pain syndrome (CP/CPPS) suffer from premature ejaculation (PE), which is difficult to cure, but the mechanism is still unknown. Based on the results of our previous clinical studies and animal experiments, we
Kun Zeng et al.
Oncotarget, 8(39), 65009-65021 (2017-10-17)
We investigated the effects of lentivirus-mediated RNAi targeting of Nogo Receptor (
Tue G Banke et al.
Nature neuroscience, 6(2), 144-152 (2003-01-14)
N-methyl-D-aspartate (NMDA) receptors are highly expressed in the central nervous system and are involved in excitatory synaptic transmission as well as synaptic plasticity. Despite considerable structural and biophysical research, the mechanism behind activation of the NMDA receptor is still poorly
W G Tingley et al.
Nature, 364(6432), 70-73 (1993-07-01)
The NMDA (N-methyl D-aspartate) receptors in the brain play a critical role in synaptic plasticity, synaptogenesis and excitotoxicity. Molecular cloning has demonstrated that NMDA receptors consist of several homologous subunits (NMDAR1, 2A-2D). A variety of studies have suggested that protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service