Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0502

Sigma-Aldrich

Gly-Ala

Synonym(s):

Glycyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH(CH3)COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1724445
EC Number:
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)CN)C(O)=O

InChI

1S/C5H10N2O3/c1-3(5(9)10)7-4(8)2-6/h3H,2,6H2,1H3,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

VPZXBVLAVMBEQI-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riccardo Chelli et al.
Journal of computational chemistry, 29(9), 1507-1516 (2008-02-14)
We have developed a fitting algorithm able to extract spectral and dynamic properties of a three level oscillator from a two-dimensional infrared spectrum (2D-IR) detected in time resolved nonlinear experiments. Such properties go from the frequencies of the ground-to-first and
Judy Tellam et al.
The Journal of experimental medicine, 204(3), 525-532 (2007-02-22)
A significant proportion of endogenously processed CD8(+) T cell epitopes are derived from newly synthesized proteins and rapidly degrading polypeptides (RDPs). It has been hypothesized that the generation of rapidly degrading polypeptides and CD8(+) T cell epitopes from these RDP
P R Tulip et al.
The Journal of chemical physics, 131(1), 015103-015103 (2009-07-10)
We investigate the structure of the glycyl-l-alanine dipeptide in aqueous solution at a 1:20 peptide:water concentration via classical, atomistic molecular dynamics simulations using the CHARMM22 force field, and compare to recent neutron diffraction data [S. E. McLain, A. K. Soper
J Rabone et al.
Science (New York, N.Y.), 329(5995), 1053-1057 (2010-08-28)
Porous materials find widespread application in storage, separation, and catalytic technologies. We report a crystalline porous solid with adaptable porosity, in which a simple dipeptide linker is arranged in a regular array by coordination to metal centers. Experiments reinforced by
Stijn Heessen et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(3), 1532-1537 (2002-01-24)
Functional inactivation of the tumor suppressor protein p53 by accelerated ubiquitin/proteasome-dependent proteolysis is a common event in tumor progression. Proteasomal degradation is inhibited by the Gly-Ala repeat (GAr) of the Epstein-Barr virus nuclear antigen-1, which acts as a transferable element

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service