Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0267

Sigma-Aldrich

Mini Gastrin I human

≥97% (HPLC)

Synonym(s):

HG-13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H99N15O26S
CAS Number:
Molecular Weight:
1646.73
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](N)CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc4ccccc4)C(N)=O

Gene Information

human ... GAST(2520)

Amino Acid Sequence

Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2

Biochem/physiol Actions

CCKB/gastrin receptor agonist. Corresponds to fragment 5-17 of human gastrin I. This form of gastrin has been isolated from tumor tissue.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth von Guggenberg et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 14(3), 366-375 (2011-07-07)
Targeting of cholecystokinin receptor expressing malignancies such as medullary thyroid carcinoma is currently limited by low in vivo stability of radioligands. To increase the stability, we have developed and preclinically evaluated two cyclic 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA)-minigastrin analogs radiolabeled with (111)In
Stephan Good et al.
European journal of nuclear medicine and molecular imaging, 35(10), 1868-1877 (2008-05-30)
Diethylenetriamine-pentaacetic acid (DTPA)-coupled minigastrins are unsuitable for therapeutic application with the available beta-emitting radiometals due to low complex stability. Low tumour-to-kidney ratio of the known radiopharmaceuticals is further limiting their potency. We used macrocyclic chelators for coupling to increase complex
Elisabeth Laabs et al.
Investigative radiology, 46(3), 196-201 (2010-12-09)
A variety of tumors in different organs with good accessibility to near-infrared light express the cholecystokinin-2 (CCK₂)/gastrin receptor. Therefore, the applicability of fluorescence optical imaging was assessed using a novel peptide probe. This study was approved by the regional animal
Anna Lucia Tornesello et al.
Journal of peptide science : an official publication of the European Peptide Society, 17(5), 405-412 (2011-02-26)
The development of suitable radioligands for targeting CCK-2 receptor expressing tumors, such as medullary thyroid carcinoma, is of great clinical interest. In the search for the best CCK-2R binding peptides, we have synthesized, evaluated and compared the CCK8 peptide (Asp-Tyr-Met-Gly-Trp-Met-Asp-PheNH(2)
Johanna Dalmo et al.
Oncology reports, 27(1), 174-181 (2011-10-14)
To be able to evaluate new radiopharmaceuticals and optimize diagnostic and therapeutic procedures, relevant animal models are required. The aim of this study was to evaluate the medullary thyroid carcinoma GOT2 animal model by analyzing the biodistribution of 177Lu-octreotate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service