Skip to Content
MilliporeSigma
All Photos(1)

Documents

F9761

Sigma-Aldrich

1-Fluoro-1-deoxy-D-fructose

~98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11FO5
CAS Number:
Molecular Weight:
182.15
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~98% (TLC)

storage temp.

2-8°C

InChI

1S/C6H11FO5/c7-1-3(9)5(11)6(12)4(10)2-8/h4-6,8,10-12H,1-2H2

InChI key

RSKQTWRWSVXAHS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service