Skip to Content
MilliporeSigma
All Photos(1)

Documents

F8931

Sigma-Aldrich

Anti-Flotillin 1-CY3 antibody produced in rabbit

1.0-3.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Flot1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

concentration

1.0-3.0 mg/mL

technique(s)

direct immunofluorescence: 2-5 μg/mL using human HEK-293T or HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLOT1(10211)
mouse ... Flot1(14251)
rat ... Flot1(64665)

General description

Flotillins are lipid raft-associated proteins belonging to the integral membrane proteins. It is associated in different biological processes such as endocytosis, phagocytosis, neuronal regeneration, insulin signaling etc. It exhibits an evolutionarily conserved domain called the prohibitin homology (PHB) domain or SPFH domain (stomatin/prohibitin/flotillin/HflK/C domain). The lipid recognition motif present in the PHB domain, is responsible for targeting proteins to the plasma membrane. Flotillin protein mainly possess two forms, flotillin 1 and 2. They are also known as reggie-2 and 1, respectively.

Immunogen

synthetic peptide corresponding to amino acid residues 413-428 of mouse flotillin 1, conjugated to KLH. The corresponding sequence is identical in rat and differs by one amino acid in human.

Application

Anti-Flotillin 1-CY3 antibody is suitable for direct immunofluorescence at a concentration of 2-5μg/mL using human HEK-293T or HeLa cells

Biochem/physiol Actions

Flotillin 1 is directly connected to the plasma membrane. The association of flotillin 1 depends on the palmitoylation in a conserved Cys34 residue, whereas the association of flotillin 2 depends on both myristolation and palmitoylation. Both the form possess a complementary tissue distribution. Flotillin 1 is mainly expressed in striated muscle tissues, adipose tissue and lung, whereas flotillin 2 is much widely found but absent from skeletal muscle. It has been reported that flotillin 1 act as a molecular link between lipid rafts of the plasma membrane and a multimeric signaling complex at the actin cytoskeleton.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Legal Information

Cy3 is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Liu et al.
The Journal of biological chemistry, 280(16), 16125-16134 (2005-02-17)
Flotillin-1 is a lipid raft-associated protein that has been implicated in various cellular processes. We examined the subcellular distribution of flotillin-1 in different cell types and found that localization is cell type-specific. Flotillin-1 relocates from a cytoplasmic compartment to the
D Volonte et al.
The Journal of biological chemistry, 274(18), 12702-12709 (1999-04-23)
Caveolae are vesicular organelles that represent a subcompartment of the plasma membrane. Caveolins and flotillins are two families of mammalian caveolae-associated integral membrane proteins. However, it remains unknown whether flotillins interact with caveolin proteins to form a stable caveolar complex
Isabel C Morrow et al.
The Journal of biological chemistry, 277(50), 48834-48841 (2002-10-09)
Flotillins are lipid raft-associated proteins, which have been implicated in neuronal regeneration and insulin signaling. We now show that newly synthesized flotillin-1 reaches the plasma membrane via a Sar1-independent and brefeldin A-resistant targeting pathway. Consistent with post-translational membrane association of
P E Bickel et al.
The Journal of biological chemistry, 272(21), 13793-13802 (1997-05-23)
Caveolae are plasmalemmal microdomains that are involved in vesicular trafficking and signal transduction. We have sought to identify novel integral membrane proteins of caveolae. Here we describe the identification and molecular cloning of flotillin. By several independent methods, flotillin behaves
Carolin Neumann-Giesen et al.
The Biochemical journal, 378(Pt 2), 509-518 (2003-11-06)
The reggie protein family consists of two proteins, reggie-1 and -2, also called flotillins, which are highly ubiquitous and evolutionarily conserved. Both reggies have been shown to be associated with membrane rafts and are involved in various cellular processes such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service